Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO6/TMEM16F Mouse anti-Human, Clone: N429/19, MilliporeSigma™

Catalog No. MABN186625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN186625 25 μL
MABN1866100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN186625 Supplier MilliporeSigma Supplier No. MABN186625UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ANO6/TMEM16F Monoclonal antibody specifically detects ANO6/TMEM16F in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANO6/TMEM16F
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone N429/19
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Recombinant fragment corresponding to the first 294 amino acids from the N-terminal, cytoplasmic domain of human Anoctamin-6 (ANO6).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.