Learn More
Invitrogen™ ANO7 Polyclonal Antibody

Description
ANO7 Polyclonal Antibody detects endogenous levels of ANO7 protein.
Specifications
Specifications
| Antigen | ANO7 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ANO7 |
| Gene Accession No. | Q6IWH7 |
| Gene Alias | ANO7; anoctamin 7; anoctamin-7; Dresden transmembrane protein of the prostate; Dresden-transmembrane protein of the prostate; DTMPP; D-TMPP; IPCA5; IPCA-5; New gene expressed in prostate; NGEP; PCANAP5; PCANAP5L; prostate cancer associated protein 5; Prostate cancer-associated protein 5; TMEM16G; Transmembrane protein 16G |
| Gene Symbols | ANO7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.