Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANO7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317120100UL
Description
ANO7 Polyclonal antibody specifically detects ANO7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ANO7 | |
Polyclonal | |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
anoctamin 7, Dresden transmembrane protein of the prostate, IPCA-5IPCA5, New gene expressed in prostate, NGEPD-TMPP, PCANAP5anoctamin-7, PCANAP5L, prostate cancer associated protein 5, prostate cancer-associated gene 5, Prostate cancer-associated protein 5, TMEM16GDresden-transmembrane protein of the prostate, Transmembrane protein 16GDTMPP | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DLLVPDIPESVEIKVKREYYLAKQALAENEVLFGTNGTKDEQPEGSELSSHWTPFTVPKASQLQQ | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
50636 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction