Learn More
Invitrogen™ ANO9 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ANO9 protein.
Specifications
Specifications
| Antigen | ANO9 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ANO9 |
| Gene Accession No. | A1A5B4 |
| Gene Alias | 5430425C04Rik; ANO9; anoctamin 9; anoctamin-9; p53-induced gene 5 protein; PIG5; RGD1560565; TM16J; TMEM16J; TP53I5; transformation related protein 53 inducible protein 5; transmembrane protein 16J; Trp53 inducible protein 5; Trp53i5; tumor protein p53 inducible protein 5; tumor protein p53-inducible protein 5 |
| Gene Symbols | ANO9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.