Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANO9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559004 Supplier Invitrogen™ Supplier No. PA559004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTTIPVTTRI RIVNFVVMNN KTSAGETFED LMKDGVFEAR FPLHKGEGRL KKTWARWRHM FREQPVDEIR NY Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 58%.

Does not exhibit calcium-activated chloride channel (CaCC) activity. Can inhibit the activity of ANO1. Jikuya H., DNA Res. 10:49-57(2003). The MGC Project Team, Genome Res. 14:2121-2127(2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANO9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANO9
Gene Accession No. A1A5B4
Gene Alias 5430425C04Rik; ANO9; anoctamin 9; anoctamin-9; p53-induced gene 5 protein; PIG5; RGD1560565; TM16J; TMEM16J; TP53I5; transformation related protein 53 inducible protein 5; transmembrane protein 16J; Trp53 inducible protein 5; Trp53i5; tumor protein p53 inducible protein 5; tumor protein p53-inducible protein 5
Gene Symbols ANO9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANO9. Recombinant protein control fragment (Product #RP-97529).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.