Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANP32A Polyclonal Antibody

Catalog No. PIPA5122238
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122238 20 μL
PIPA5118925 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122238 Supplier Invitrogen™ Supplier No. PA5122238
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANP32A Polyclonal Antibody for IHC (P), Western Blot

PHAP I is implicated in a number of cellular processes, including proliferation, differentiation, caspase-dependent and caspase-independent apoptosis, suppression of transformation (tumor suppressor), inhibition of protein phosphatase 2A, regulation of mRNA trafficking and stability in association with ELAVL1, and inhibition of acetyltransferases as part of the INHAT (inhibitor of histone acetyltransferases) complex. Plays a role in E4F1-mediated transcriptional repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANP32A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ANP32A
Gene Accession No. P39687, P49911
Gene Alias Acid nuclear phosphoprotein 32 (leucine rich); acidic (leucine-rich) nuclear phosphoprotein 32 family, member A; acidic leucine-rich nuclear phosphoprotein 32 family member A; acidic nuclear phosphoprotein 32; acidic nuclear phosphoprotein 32 family member A; acidic nuclear phosphoprotein 32 family, member A; acidic nuclear phosphoprotein pp32; Anp32; ANP32A; C15orf1; cerebellar leucine rich acidic nuclear protein; hepatopoietin Cn; HPPCn; I1PP2A; inhibitor-1 of protein phosphatase-2A; LANP; Leucine-rich acidic nuclear protein; leucine-rich acidic nuclear protein LANP; MAPM; Mapmodulin; MGC119787; MGC150373; PHAP1; PHAPI; Potent heat-stable protein phosphatase 2A inhibitor I1PP2A; pp32; Putative HLA-DR-associated protein I; putative human HLA class II associated protein I; vacidic (leucine-rich) nuclear phosphoprotein 32 family, member A; W91701
Gene Symbols ANP32A
Host Species Rabbit
Immunogen Recombinant protein Acidic Nuclear Phosphoprotein 32 Family, Member A. The antigen corresponds to amino acid range 2-175 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25379, 8125
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.