Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANP32B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | ANP32B |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2-4 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056369T
![]() |
Novus Biologicals
NB10056369SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056369
![]() |
Novus Biologicals
NB10056369 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
ANP32B Polyclonal specifically detects ANP32B in Human samples. It is validated for Western Blot.Specifications
ANP32B | |
Western Blot 2-4 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.02% Sodium Azide | |
acidic (leucine-rich) nuclear phosphoprotein 32 family, member B, acidic leucine-rich nuclear phosphoprotein 32 family member B, Acidic protein rich in leucines, APRILSilver-stainable protein SSP29, Pal31, PHAPI2SSP29, Putative HLA-DR-associated protein I-2 | |
ANP32B | |
IgG | |
Protein G purified |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
NP_003799.1 | |
10541 | |
This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 1-17 of human APRIL. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title