Learn More
Invitrogen™ ANP32C Monoclonal Antibody (OTI10A10)

Description
ANP32C Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ANP32C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI10A10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ANP32C |
| Gene Accession No. | O43423 |
| Gene Alias | acidic (leucine-rich) nuclear phosphoprotein 32 family, member C; acidic (leucine-rich) nuclear phosphoprotein 32 family, pseudogene; acidic leucine-rich nuclear phosphoprotein 32 family member C; acidic nuclear phosphoprotein 32 family member C; ANP32C; Anp32cl; Anp32c-ps; Anp32-ps; EG448829; Phosphoprotein 32-related protein 1; pp32 related 1; PP32R1; Tumorigenic protein pp32r1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.