Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANP32D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103290 Supplier Invitrogen™ Supplier No. PA5103290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ANP32D.

ANP32D (acidic (leucine-rich) nuclear phosphoprotein 32 family, member D), also known as PP32R2 (phosphoprotein 32-related protein 2) or tumorigenic protein pp32r2, is a 131 amino acid protein that contains 3 leucine-rich repeats and belongs to the ANP32 family. ANP32D shares 89.3% amino acid sequence identity with ANP32A, a tumor suppressor that inhibits a variety of cancers, such as prostate and breast cancers. Although similar to ANP32A, ANP32D is tumorigenic and is preferentially expressed in prostatic cancer. Conversely, it is uncertain that ANP32D is neurally expressed. Intronless, ANP32D may be a pseudogene that is expressed in pathological situations and may also modulate the oncogenic potential of human prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANP32D
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ANP32D
Gene Accession No. O95626
Gene Alias acidic (leucine-rich) nuclear phosphoprotein 32 family, member D; acidic leucine-rich nuclear phosphoprotein 32 family member D; acidic nuclear phosphoprotein 32 family member D; acidic nuclear phosphoprotein 32D; ANP32D; Phosphoprotein 32-related protein 2; pp32 related 2; PP32R2; Tumorigenic protein pp32r2
Gene Symbols ANP32D
Host Species Rabbit
Immunogen A synthesized peptide derived from human ANP32D(Accession O95626), corresponding to amino acid residues M1-E40.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23519
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.