Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANPEP Mouse anti-Human, Unconjugated, Clone: APN/514, Abnova™

Catalog No. 89337523 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-523 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-523 Supplier Abnova Corporation Supplier No. MAB21030
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ANPEP Monoclonal antibody specifically detects ANPEP in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen ANPEP
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone APN/514
Conjugate Unconjugated
Description alanyl (membrane) aminopeptidase
Gene ANPEP
Gene Alias APN, CD13, LAP1, PEPN, gp150, p150
Host Species Mouse
Immunogen Recombinant protein corresponding to human ANPEP
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 290
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.