Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANPEP Rabbit anti-Human, Clone:RM314, Abnova™

Catalog No. 89333295 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-295 Supplier Abnova Corporation Supplier No. MAB16939
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against huamn ANPEP.

Aminopeptidase N is located in the small-intestinal and renal microvillar membrane, and also in other plasma membranes. In the small intestine aminopeptidase N plays a role in the final digestion of peptides generated from hydrolysis of proteins by gastric and pancreatic proteases. Its function in proximal tubular epithelial cells and other cell types is less clear. The large extracellular carboxyterminal domain contains a pentapeptide consensus sequence characteristic of members of the zinc-binding metalloproteinase superfamily. Sequence comparisons with known enzymes of this class showed that CD13 and aminopeptidase N are identical. The latter enzyme was thought to be involved in the metabolism of regulatory peptides by diverse cell types, including small intestinal and renal tubular epithelial cells, macrophages, granulocytes, and synaptic membranes from the CNS. Human aminopeptidase N is a receptor for one strain of human coronavirus that is an important cause of upper respiratory tract infections. Defects in this gene appear to be a cause of various types of leukemia or lymphoma. [provided by RefSeq]

Specifications

Antigen ANPEP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RM314
Conjugate Unconjugated
Description alanyl (membrane) aminopeptidase
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1000)
Western Blot (1:1000-2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene ANPEP
Gene Alias APN, CD13, LAP1, PEPN, gp150, p150
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human ANPEP.
Quantity 100 μL
Gene ID (Entrez) 290
Test Specificity This antibody reacts to human CD5.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.