Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Anpirtoline hydrochloride
GREENER_CHOICESDP

Catalog No. 070350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-035-0 50 mg
07-031-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-035-0 Supplier R&D Systems™ Supplier No. 0703/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent 5-HT1B agonist; also 5-HT3 antagonist

Highly potent 5-HT1B receptor agonist (Ki values are 28, 150 and 1490 nM at 5-HT1B, 5-HT1A and 5-HT2 receptors respectively). Decreases central serotonin synthesis and attenuates aggressive behavior in vivo. Also acts as an antagonist at 5-HT3 receptors (Ki = 29.5 nM) and is brain penetrant.

Chemical Identifiers

CAS 99201-87-3
Molecular Formula C10H14Cl2N2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 265.2
Purity >99%
Molecular Weight (g/mol) 265.196
Chemical Name or Material Anpirtoline hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.