Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANR44 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566022
Description
Immunogen sequence: ASIFVKDNVTK RTPLHASVIN GHTLCLRLLL EIADNPEAVD VKDAKGQTP Highest antigen sequence identity to the following orthologs - mouse 96%, rat 98%.
Putative regulatory subunit of protein phospatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates.
Specifications
ANR44 | |
Polyclonal | |
Unconjugated | |
ANKRD44 | |
ANKRD44; ankyrin repeat domain 44; ankyrin repeat domain-containing protein 44; PP6-ARS-B; protein phosphatase 6 ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory subunit ARS-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N8A2 | |
ANKRD44 | |
Recombinant Human ANR44. Recombinant protein control fragment (Product #RP-106712). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction