Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANR44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597101 Supplier Invitrogen™ Supplier No. PA597101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHMTAVHGRF TRSQTLIQNG GEIDCVDKDG NTPLHVAARY GHELLINTLI TSGADTAKCG IHSMFPLHLA ALNAHSDCCR KLLSSGFEID TPDKFGRTCL HAAAAGGNVE CIKLLQSSGA DFHKKDKCGR TPLHYAAANC HFHCIETLVT TGANVNETDD WGRTALHYAA ASDMDRNKTI LGNAHDNSEE LERARELKEK EATLCLEFLL QNDANPSIRD KEGYNSIHYA AAYGHRQCLE L; Positive Samples: H460, Jurkat.

Putative regulatory subunit of protein phospatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANR44
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANKRD44
Gene Accession No. Q8N8A2
Gene Alias ANKRD44; ankyrin repeat domain 44; ankyrin repeat domain-containing protein 44; PP6-ARS-B; protein phosphatase 6 ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B; serine/threonine-protein phosphatase 6 regulatory subunit ARS-B
Gene Symbols ANKRD44
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 310-550 of human ANKRD44 (NP_0011820731).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91526
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.