Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANR49 Monoclonal Antibody (OTI3E4)
GREENER_CHOICE

Catalog No. PIMA526333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526333 Supplier Invitrogen™ Supplier No. MA526333
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ANR49 Monoclonal Antibody for Western Blot, ICC/IF

Induces HBG1 expression (PubMed:16131492, PubMed:11162141). May have a role in spermatogenesis where it promotes autophagy in response to serum starvation, via the NF-kappaB pathway. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANR49
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANKRD49
Gene Accession No. Q8WVL7
Gene Alias ANKRD49; ankyrin repeat domain 49; ankyrin repeat domain-containing protein 49; AW060934; C86108; fetal globin inducing factor; fetal globin-increasing factor; Fetal globin-inducing factor; FGIF; Gbif; globin inducing factor, fetal
Gene Symbols ANKRD49
Host Species Mouse
Immunogen Full length human recombinant protein of ANKRD49 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54851
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.