Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANR49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559131 Supplier Invitrogen™ Supplier No. PA559131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLAAGNRDSK DTLELLLMNR YVKPGLKNNL EETAFDIARR TSIYHYLFEI VEGCTNSSPQ Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

Induces HBG1 expression (PubMed:16131492, PubMed:11162141). May have a role in spermatogenesis where it promotes autophagy in response to serum starvation, via the NF-kappaB pathway. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANR49
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD49
Gene Accession No. Q8WVL7
Gene Alias ANKRD49; ankyrin repeat domain 49; ankyrin repeat domain-containing protein 49; AW060934; C86108; fetal globin inducing factor; fetal globin-increasing factor; Fetal globin-inducing factor; FGIF; Gbif; globin inducing factor, fetal
Gene Symbols ANKRD49
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANR49. Recombinant protein control fragment (Product #RP-97465).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.