Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANR52 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560523
Description
Immunogen sequence: HYAAASDTYR RAEPHTPSSH DAEEDEPLKE SRRKEAFFCL EFLLDNG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. [UniProt].
Specifications
ANR52 | |
Polyclonal | |
Unconjugated | |
ANKRD52 | |
6430544C07; ANKRD33; ANKRD52; ankyrin repeat domain 33; ankyrin repeat domain 52; ankyrin repeat domain-containing protein 52; CVWG5837; G431002C21Rik; PP6-ARS-C; protein phosphatase 6 ankyrin repeat subunit C; RGD1307124; Serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C; serine/threonine-protein phosphatase 6 regulatory subunit ARS-C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
283373 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NB46 | |
ANKRD52 | |
Recombinant protein corresponding to Human ANR52. Recombinant protein control fragment (Product #RP-100403). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction