Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANR52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121191 Supplier Invitrogen™ Supplier No. PA5121191
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-251MG, HeLa, A-549, K-562, BT-474, Mouse brain, Mouse liver, Rat brain, Rat liver. Immunogen sequence: RRRDKFGRTP LHYAAANGSY QCAVTLVTAG AGVNEADCKG CSPLHYAAAS DTYRRAEPHT PSSHDAEEDE PLKESRRKEA FFCLEFLLDN GADPSLRDRQ GYTAVHYAAA YGNRQNLELL LEMSFNCLED VESTIPVSPL HLAAYNGHCE ALKTLAETLV NLDVRDHKGR TALFLATERG STECVEVLTA HGASALIKER KRKWTPLHAA A.

Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANR52
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANKRD52
Gene Accession No. Q8BTI7, Q8NB46
Gene Alias 6430544C07; ANKRD33; ANKRD52; ankyrin repeat domain 33; ankyrin repeat domain 52; ankyrin repeat domain-containing protein 52; CVWG5837; G431002C21Rik; PP6-ARS-C; protein phosphatase 6 ankyrin repeat subunit C; RGD1307124; Serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C; serine/threonine-protein phosphatase 6 regulatory subunit ARS-C
Gene Symbols ANKRD52
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 450-660 of human ANKRD52 (NP_775866.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237615, 283373, 362811
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.