Ansell™ HyFlex™ Ultralight Weight Multi-purpose Gloves
Gloves feature ZONZ™ Comfort Fit technology
Manufacturer: Ansell™ 11818 7 111714
 View more versions of this product
Catalog No. 19-148-411
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Assembly of dry parts, feeding lines, fine mounting of small nuts and bolts, Handling of small/fine parts, Mounting, dismounting parts, Screwing & unscrewing, ultra-fine mechanical applications
    Length (Metric) 200-248mm
    Size 7
    Color Dark blue
  • Description & Specifications
Related Products
Product Content Feedback