Learn More
Ansell™ Sawyer-Tower CPC Nomex Jacket
Catalog No. 19160395E Shop All Ansell Products
Change view
Click to view available options
Blue CPC Nomex Jacket; Size: XX-Large
Catalog No.
19-160-395E Blue CPC Nomex Jacket; Size: XX-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-160-395E Supplier Ansell™ Supplier No. 666702XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Breathable chemical splash protection
  • Gore-Tex fabric offers exceptional comfort and outstanding chemical splash protection Breathability of the Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
  • Protects workers from certain liquid and dry chemical exposures
  • Lightweight, flexible, snag- and tear-resistant, stain-repellent, and washable
TRUSTED_SUSTAINABILITY
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.