Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ TRELLCHEM™ ACT Coveralls with Bayonet glove ring system

Catalog No. 19010069 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-069 3X-Large
19-010-064 Small
19-010-065 Medium
19-010-066 Large
19-010-067 X-Large
19-010-068 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-010-069 Supplier Ansell™ Supplier No. 164206
This item is not returnable. View return policy

Requires

Onguard Hazmax™ boots and SCBA units by Scott and MSA

Specially designed for protection in CBRN terrorism incidents and IDLH

  • Made of techinical barrier fabric with a fluoro-based barrier film
  • Non-encapsulating design (Level B/TypeT) with mask seal and without hump, self-contained breathing apparatus (SCBA) worn outside the suit
  • Elastic waist and shoulder paddings for added comfort
  • Attached hood with 3D shaped butyl rubber mask seal
  • long zipper positioned diagonally over the chest, closing down-wards for added safety. The zipper is protected by a double splash guard(flap)
  • Bayonet glove ring system, which offers quick and simple glove replacement
  • Equipped with sewn-in sock/bootie in suit material
  • Seams are stitched with aramid thread sealed by a welded-on barrier strip, seamless barrier and optimal safety
  • Available in XS to XXXL sizes
  • Black in color

Optional Features:

  • External pockets on chest, waist and/or hip
  • Paddings on knees and elbows for improved comfort and safety
  • Reflective markings on sleeves and legs for high visibilitybullets1 (end)
Applications:
  • Law enforcement
  • First responders
  • Hazmat teams
  • Hot Zonesearch and rescue
  • Explosive Ordnance Disposal (EOD)
  • Clandestine lab entry, drug raids
  • Technical rescue in contaminated environment
  • Decontamination
  • Tactical operations: Critical infrastucture protection, Quick response forces
  • CBRNE response
  • Civil Support Teams: USAR teams, Triage, Medical operations

Compliance

American standard NFPA1994 (class2)

TRUSTED_SUSTAINABILITY

Specifications

Size 3X-Large
Material Technical Barrier Fabric
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.