Description
- Made of techinical barrier fabric with a fluoro-based barrier film
- Non-encapsulating design (Level B/TypeT) with mask seal and without hump, self-contained breathing apparatus (SCBA) worn outside the suit
- Elastic waist and shoulder paddings for added comfort
- Attached hood with 3D shaped butyl rubber mask seal
- long zipper positioned diagonally over the chest, closing down-wards for added safety. The zipper is protected by a double splash guard(flap)
- Glove attachment: 5-finger butyl rubber glove with TRELLCHEM ACT Overglove for extra protection
- Equipped with sewn-in sock/bootie in suit material
- Seams are stitched with aramid thread sealed by a welded-on barrier strip, seamless barrier and optimal safety
- Available in XS to XXXL sizes
- Black in color
Optional Features:
- External pockets on chest, waist and/or hip
- Paddings on knees and elbows for improved comfort and safety
- Reflective markings on sleeves and legs for high visibility
- Law enforcement
- First responders
- Hazmat teams
- Hot Zonesearch and rescue
- Explosive Ordnance Disposal (EOD)
- Clandestine lab entry, drug raids
- Technical rescue in contaminated environment
- Decontamination
- Tactical operations: Critical infrastucture protection, Quick response forces
- CBRNE response
- Civil Support Teams: USAR teams, Triage, Medical operations
Specifications
Specifications
| Size | Large |
| Material | Technical Barrier Fabric |
| Color | Black |