Learn More
Ansell™ TRELLCHEM™ VPS-Flash Coveralls
Catalog No. p-5237627
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Protects against hazardous chemicals in liquid, vapor, gaseous and solid form, including warfare agents
  • Made of combination of elastomers and plastics with a No-mex™ fabric
  • Chemical, flame and abrasion resistant
  • Available in both encapsulating and non-encapsulating designs
  • Option of two visors; CV or the larger VP1 made of PVC
  • Gastight chloroprene rubber coated zipper
  • Trellchem™ regulating valve has 3 ventilation rates (2, 30 and 100 l/min) plus zero/off position 
  • Glove attachments: Inner Ansell Barrier and Trellchem™ Viton™ /Butyl rubber glove
  • AlphaTec overglove for extra cut protection
  • Glove are attached with Trellchem™ Bayonet glove ring system
  • Equipped with sewn-on sock/bootie and alternatively suit has attached boots 
  • Stitched with aramid thread, taped with rubber strip on outside and barrier laminate strip welded inside
Compliance
Meets NFPA 1991 Standard on Vapor Protective Ensembles for Hazardous Materials Emergencies and CBRN Terrorism Incident.
TRUSTED_SUSTAINABILITY
Specifications
Color Orange, Black
Material Nomex™
Size X-Small
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.