Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ANSI Type R Class 2 Expandable Two-Tone Mesh Breakaway Vest

Catalog No. 19046485
Click to view available options
Size:
2X-Large to 5X-Large
Medium to X-Large

Type R/Class 2 apparel is necessary for workers exposed to traffic traveling over 25 mph and those who work against complex backgrounds. Ideal for construction, municipalities, shipyards, or anywhere that hi-vis apparel is required.

  • Breathable polyester mesh with 5 point breakaway
  • Off shoulder breakaway system provides additional safety around extreme traffic hazards, or while moving machinery and equipment
  • Adjustable hook & loop sides for a universal fit
  • 2 in. two-tone tape for optimum visibility
  • Breakaway zipper front closure
  • 1 outer chest radio/cell pocket with ID pocket
  • 1 inner chest pocket
  • 1 lower inner pocket

Specifications

Certifications/Compliance ANSI/ISEA 107-2015 Class 2 Type R
Construction 5 Point Breakaway/Expandable
Closure Type Zipper
Color Hi-vis Yellow
Size 2X-Large to 5X-Large
Material Polyester Mesh
Length (English) 28 in.
Length (Metric) 71 cm
Size (English) Chest 57.9 in.
No. of Pockets 3
Size (Metric) Chest 147 cm
Tape Type Two-Tone
For Use With (Application) Construction, Surveying, Shipyards, and Municipalities
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.