Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ANSI Type R Class 2 Three Pocket Mesh Breakaway Vest

Catalog No. 19046481 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-481 4X-Large
19-046-475 Medium
19-046-476 Large
19-046-478 X-Large
19-046-479 2X-Large
19-046-480 3X-Large
19-046-482 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-046-481 Supplier PIP™ Supplier No. 3025PMVLY4X
Add to Cart
Add to Cart

Type R/Class 2 apparel is necessary for workers exposed to traffic traveling over 25 mph and those who work against complex backgrounds. Ideal for construction, municipalities, shipyards, or anywhere that hi-vis apparel is required.

  • Breathable polyester mesh
  • Breakaway provides additional safety around extreme traffic hazards, or moving machinery and equipment
  • Hook & Loop closure
  • 2 in. silver reflective tape
  • 1 external chest pocket with 1 pen pocket
  • 2 internal lower pockets

Specifications

Product Type High-visibility Vest
Certifications/Compliance ANSI/ISEA 107-2015 Class 3 Type R
Construction 5 Point Breakaway
Closure Type Hook and Loop
Color Hi-vis Yellow
Size 4X-Large
Material Polyester Mesh
Length (English) 30 in.
Length (Metric) 76 cm
Size (English) Chest 59 in.
No. of Pockets 3
Size (Metric) Chest 150 cm
Tape Type Level 2 Silver Reflective (2 in.)
For Use With (Application) Construction, Municipalities, Shipyards, and Railroads
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.