Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANT Recombinant Rabbit Monoclonal Antibody (3C1Q3)
GREENER_CHOICE

Catalog No. FEMA551279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551279 Supplier Invitrogen™ Supplier No. MA551279
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ANT Recombinant Monoclonal Antibody for ELISA, ICC/IF, IP, Western Blot

ANT1 catalyzes the exchange of ADP and ATP across the mitochondrial inner membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANT
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3C1Q3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC25A4
Gene Accession No. P12235, P48962, Q05962
Gene Alias 30 kDa calsequestrin-binding protein; 30 kDa CSQ-binding protein; AAC1; adenine nucleotide translocase-1; adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; ADP ATP translocase; ADP,ATP carrier protein 1; ADP,ATP carrier protein, heart; ADP,ATP carrier protein, heart isoform T1; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP carrier; ADP/ATP translocase; ADP/ATP translocase 1; ADP/ATP translocase T1; ADP/ATP-carrier protein; Anc1; ANT; ANT 1; ANT1; AU019225; CSQ-binding 30 kDa protein; fa22e07; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; MTDPS12; PEO2; PEO3; PEOA2; Slc25a4; SLC25A5; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; solute carrier family 25 member 4; T1; translocase; Unknown (protein for MGC:77591); wu:fa22e07; zgc:77591
Gene Symbols SLC25A4
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11739, 291, 85333
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.