Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Antacid Effervescent
SCI_ED

Catalog No. s25180 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
24 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25180 24 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25180
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains sodium bicarbonate (50%), citric acid (30-40%) amd acetylsalicylic acid (10%)

  • Non-flammable
  • Effervesces in water

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Product Type Antacid Effervescent Tablet
Active Ingredients Sodium Bicarbonate (50%), Citric Acid (30 to 40%), Acetylsalicylic Acid (10%), Calcium Carbonate (40%, w/w)
Format Tablet
Quantity 24 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.