Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antennapedia Control Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7110689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223629SS 0.025 mg
NBP223629 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223629SS Supplier Novus Biologicals Supplier No. NBP223629SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antennapedia Control Polyclonal specifically detects Antennapedia Control in Mouse, All Species samples. It is validated for Western Blot.

Specifications

Antigen Antennapedia Control
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Antennapedia homeodomain
Gene Alias AP homeodomain, AP homeodomain Drosophilia
Host Species Rabbit
Immunogen Reacts with the Antennapedia homeodomain sequence RQIKIWFQNRRMKWKK, which is 100 % conserved across all known species.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.