Alfa Aesar™ Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%

Manufacturer: Alfa Aesar™ H5192103

Quantity 1g

 View more versions of this product

Catalog No. AAH5192103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 863992-56-7
    Molecular Formula C26H32B2O4
    Formula Weight 430.16
    MDL Number MFCD16294538
    Chemical Name or Material Anthracene-9,10-diboronic acid bis(pinacol) ester

    Specifications

    Formula Weight 430.16
    Molecular Formula C26H32B2O4
    MDL Number MFCD16294538
    CAS 863992-56-7
    Quantity 1g
    Chemical Name or Material Anthracene-9,10-diboronic acid bis(pinacol) ester
    Assay 95%

    Description

    Applications
    It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n ft.-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit