Alfa Aesar™ Anthracene, 99%

Manufacturer: Alfa Aesar™ A1020314

Quantity 25g

 View more versions of this product

Catalog No. AAA1020314

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 120-12-7
    Molecular Formula C14H10
    Formula Weight 178.23
    MDL Number MFCD00001240
    UN Number UN3077
    Beilstein 1905429
    Merck Index 14,682
    Chemical Name or Material Anthracene

    Specifications

    Formula Weight 178.23
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3077
    Beilstein 1905429
    Health Hazard 1 Warning
    Boiling Point 340° to 342°C
    Quantity 25g
    Flash Point 121°C (249°F)
    TSCA TSCA
    Molecular Formula C14H10
    CAS 120-12-7
    Merck Index 14,682
    Melting Point 214° to 218°C
    RTECS Number CA9350000
    MDL Number MFCD00001240
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Anthracene
    Assay 99%

    Description

    Applications
    Anthracene is used in the production of the red dyealizarin and other dyes.

    Solubility
    Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit