Learn More
Eisco™ Anthracite Coal Specimen
SCI_ED
Catalog No. S24777 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24777 12/Pk. 2 to 3 cm
S24776 1 Ea. 2 to 3 cm
S26610 10/Pk. 2 to 3 cm
S26663 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24777 Supplier Eisco™ Supplier No. ESNG0053PK12
Add to Cart
Add to Cart
Anthracite Coal is formed in anoxic environments from compressed organic matter, and is used worldwide as fuel source.
  • Denser than bituminous coal, with higher proportion of carbon
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Closer look at material that powers world
  • Bright and reflective, indicative of intense heat and pressure required to form it
Specifications
Product Type Metamorphic Rock
For Use With (Application) Classroom identification labs and exercises
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Anthracite Coal