Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Anthracite Coal Specimen
SCI_ED

Catalog No. S26663 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S26663 1 Ea. 7 to 8 cm
S24776 1 Ea. 2 to 3 cm
S26610 10/Pk. 2 to 3 cm
S24777 12/Pk. 2 to 3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26663 Supplier Eisco™ Supplier No. ESNG0053HAND
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anthracite Coal is formed in anoxic environments from compressed organic matter, and is used worldwide as fuel source.

  • Denser than bituminous coal, with higher proportion of carbon
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Closer look at material that powers world
  • Bright and reflective, indicative of intense heat and pressure required to form it

Specifications

Product Type Metamorphic Rock
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Specimen Type Anthracite Coal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.