Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anthrax EF Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
| Antigen | Anthrax EF |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Anthrax EF Polyclonal specifically detects Anthrax EF in Bacteria samples. It is validated for ELISA.Specifications
| Anthrax EF | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide corresponding to 16 amino acids near the carboxy terminus of the Anthrax edema factor protein. The immunogen is located within the last 50 amino acids of Anthrax Edema Factor. Amino Acid Squence: NFTENETDNFEVFQK | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Bacteria | |
| Adenylyl cyclase, Anthrax edema toxin adenylate cyclase component, ATP pyrophosphate-lyase, Calmodulin-sensitive adenylate cyclase, Edema factor 1 (EF 1) | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title