Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anthrax LF Antibody (5F502.2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056592H
Description
Anthrax LF Monoclonal specifically detects Anthrax LF in Human samples. It is validated for Western Blot.Specifications
Anthrax LF | |
Monoclonal | |
HRP | |
Anthrax lethal factor, Anthrax lethal toxin endopeptidase component, Anthrax LF, B.anthracis lethal factor, bacillus anthracis lethal factor, Lef, LF | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot | |
5F502.2 | |
Western Blot | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction