Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrax LF Antibody (5F502.2), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen Anthrax LF
Clone 5F502.2
Concentration 0.5 mg/mL
Dilution Western Blot 0.1-1 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056592T
SDP
View Documents
Novus Biologicals
NB10056592SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056592
SDP
View Documents
Novus Biologicals
NB10056592
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Anthrax LF Monoclonal specifically detects Anthrax LF in Human samples. It is validated for Western Blot.
Specifications

Specifications

Anthrax LF
0.5 mg/mL
Western Blot
Unconjugated
Mouse
Human
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
5F502.2
Western Blot 0.1-1 ug/ml
Monoclonal
Purified
RUO
Anthrax lethal factor, Anthrax lethal toxin endopeptidase component, Anthrax LF, B.anthracis lethal factor, bacillus anthracis lethal factor, Lef, LF
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.