Learn More
Anthrone, 95%
Description
It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.
Solubility
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 153°C to 160°C |
| Quantity | 100 g |
| Beilstein | 1910173 |
| Sensitivity | Light sensitive |
| Merck Index | 14,691 |
| Solubility Information | Soluble in alcohol, benzene, and hot NaOH solutions; Insoluble in water |
| IUPAC Name | 9,10-dihydroanthracen-9-one |
| Formula Weight | 194.23 |
| Percent Purity | 97% |
| Physical Form | Crystalline Powder |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 201-994-0 |
| RTECSNumber | CB8925500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.