Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrone, ACS

Catalog No. AA3073922
Click to view available options
Quantity:
25 g
100 g

Used for determination of carbohydrates | CAS: 90-44-8 | C14H10O | 194.23 g/mol

Used for determination of carbohydrates Anthrone is used for the determination of carbohydrates. It is used as a general reagent in the colorimetric determination of carbohydrates in yeast and plant extracts. It is involved in organic synthesis. It is utilized for the determination of sugar in body fluids. It finds application as a laxative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
PubChem CID 7018
ChEBI CHEBI:33835
IUPAC Name 10H-anthracen-9-one
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Melting Point 155°C to 158°C
Quantity 100 g
Beilstein 1910173
Sensitivity Light sensitive
Merck Index 14,691
Solubility Information Soluble in acetone,hot benzene,concentrated sulfuric acid,hot dilute alkali and most organic solvents. Insoluble in water.
Formula Weight 194.23
Grade ACS
Physical Form Crystalline
Chemical Name or Material Anthrone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-994-0
RTECSNumber CB8925500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.