Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthrone, ACS, Spectrum™ Chemical
SDP

Catalog No. 18608892 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-892 25 g Amber Glass Bottle
18-608-893 5 g Amber Glass Bottle
18-608-891 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-608-892 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AN13525GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90-44-8

Specifications

CAS Max % 1
Melting Point Within 5°C
Packaging Amber Glass Bottle
Quantity 25 g
Grade ACS
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.