Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-1,4-N-Acetylglucosaminyltransferase 4/A4GNT Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF8555
This item is not returnable.
View return policy
Description
A4GNT Polyclonal specifically detects A4GNT in Human samples. It is validated for Western Blot.Specifications
A4GNT | |
Polyclonal | |
Western Blot 1:1000 dilution | |
Q9UNA3 | |
A4GNT | |
Chinese hamster ovary cell line CHO-derived recombinant human alpha-1,4-N-Acetylglucosaminyltransferase 4/A4GNT Leu25-Lys340 Accession # Q9UNA3 | |
100 μL | |
Primary | |
Detects human alpha-1,4-N-Acetylglucosaminyltransferase in direct ELISAs and Western blots. | |
12 months from date of receipt, -20 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 degreesC under sterile conditions after opening. | |
IgG |
Western Blot | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. with Sodium Azide | |
alpha-1,4-N-acetylglucosaminyltransferase, alpha4GnT | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
51146 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction