Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-cys Peroxiredoxin Mouse anti-Human, Rat, Clone: 8H11, MilliporeSigma™

Catalog No. MAB3478MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3478MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3478MI Supplier MilliporeSigma Supplier No. MAB3478
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect 1-cys Peroxiredoxin using this Anti-1-cys Peroxiredoxin Antibody, clone 8H11 validated for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 1-cys Peroxiredoxin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8H11
Conjugate Unconjugated
Formulation Ascites fluid. Liquid.
Gene Accession No. NM_004905.2
Gene Symbols PRDX6, p29, Peroxiredoxin-6, MGC46173, PRX, aiPLA2, AOP2, 1-Cys, KIAA0106, NSGPx, EC 1.11.1.15, EC 3.1.1.-, EC 1.11.1.7
Host Species Mouse
Immunogen Recombinant human 1-Cys peroxiredoxin.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.