Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-formyltetrahydrofolate dehydrogenase Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. ABN57MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN57MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN57MI Supplier MilliporeSigma Supplier No. ABN57

Rabbit Polyclonal Antibody

Anti-10-formyltetrahydrofolate dehydrogenase Antibody is an antibody against 10-formyltetrahydrofolate dehydrogenase for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 10-formyltetrahydrofolate dehydrogenase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Symbols 10-fTHF; FTHFD; 10-FTHFDH
Host Species Rabbit
Immunogen Recombinant protein corresponding to rat 10-formyltetrahydrofolate dehydrogenase.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_071992
Target Species Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.