Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
12L HETE, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA131514
Description
12-Hydroxyeicosatetraenoic acid, a metabolite of arachidonic acid, is a highly selective ligand used to label mu-opiod receptors in membranes and tissues. 5-Lipoxygenase (LO)-derived leukotrienes are involved in inflammatory glomerular injury. LO product 12-HETE is associated with pathogenesis of hypertension, and may mediate angiotensin II and TGF beta induced mesengial cell abnormality in diabetic nephropathy. 12-HETE is markedly elevated in the psoriatic lesions and acts as a vasoconstrictor that contributes to high blood pressure in (renovascular) hypertension and pregnancy-induced hypertension. A significant percentage of patients suffering from a selective increase in plasma LDL cholesterol (type IIa hyperlipoproteinaemia) exhibits increased platelet reactivity. This includes enhanced platelet responsiveness against a variety of platelet-stimulating agents ex vivo and enhanced arachidonic acid metabolism associated with increased generation of arachidonic acid metabolites such as 12-HETE, and secretion of platelet-storage products. The 12-s-HETE analog has been reported to augment tumour cell metastatic potential through activation of protein kinase c. 12-HETE can be formed either in the 12-S or 12-R configuration. It has a diversity of biological actions and is generated by a number of tissues including the renal glomerulus and the vasculature. 12-HETE is one of the six monohydroxy fatty acids produced by the non-enzymatic oxidation of arachidonic acid. 12-HETE is a neuromodulator that is synthesized during ischemia. Its neuronal effects include attenuation of calcium influx and glutamate release as well as inhibition of AMPA receptor (AMPA-R) activation. [National Center for Biotechnology Information. PubChem Compound Database; CID=5312983] PA1-31514 detects 12L HETE from human samples.PA1-31514 has been successfully used in radioimmune assay applications.The PA1-31514 immunogen is: Pure 12(L)-HETE conjugated to human serum albumin
Specifications
| 12L HETE | |
| Polyclonal | |
| 12-HETE, 12-Hydroxyeicosatetraenoic acid, 12(L)-Hydroxeicosantetraenoic Acid, 12(L)-HETE | |
| Pure 12(L)-HETE conjugated to human serum albumin | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Store at -20°C, Avoid Freeze/Thaw Cycles | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction