Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 Mouse, Unlabeled, Clone: 12/14-3-3, BD

Catalog No. BDB561466 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561466 150 μg
BDB561465 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561466 Supplier BD Biosciences Supplier No. 561466

Mouse Monoclonal Antibody

The family of 14-3-3 proteins has been implicated in numerous cellular processes such as neurotransmission, phospholipase activity, and calcium signaling. The 14-3-3 protein can associate with several proteins involved in cell proliferation, such as Raf-1, Bcr-Abl, middle T antigen, and CDC25 phosphatase. It appears that 14-3-3 proteins enhance the kinase activity of the Raf proto-oncogene equally as well as Ras, thus, implicating 14-3-3 proteins in the Raf/Ras/Map kinase signaling pathway. However, in spite of 14-3-3 direct association with the CDC25 phosphatase, this interaction does not increase the phosphatase activity. It has been reported that this antibody detects 14-3-3 subunits β, γ, η, Θ, and ζ , but not ε nor σ.

TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 12/14-3-3
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human 14-3-3
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgM
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.