Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 Β Rabbit anti-Bovine, Human, Mouse, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB9730
Description
Detect 14-3-3 β using this Anti-14-3-3 β Antibody validated for use in ELISA, western blot, immunohistochemistry.
Specifications
14-3-3 β | |
Polyclonal | |
Liquid. | |
Rabbit | |
Serum | |
Primary | |
Bovine, Human, Mouse, Rabbit, Rat, Sheep | |
Serum |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
YWHAB; KCIP-1; HS1; GW128 | |
Synthetic peptide from the N-terminal area of sheep 14-3-3 beta. | |
100 μL | |
NM_003404.3; NM_139323.2 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction