Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

19S regulator non-ATPase subunit S5a/Rpn10 Mouse anti-Human, Clone: S5a-18, MilliporeSigma™

Catalog No. 04259MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-259-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-259-MI Supplier MilliporeSigma Supplier No. 04259
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect 19S regulator non-ATPase subunit S5a/Rpn10 using this Anti-19S regulator non-ATPase subunit S5a/Rpn10 Antibody, clone S5a-18 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 19S regulator non-ATPase subunit S5a/Rpn10
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S5a-18
Conjugate Unconjugated
Formulation 100μL of purified immunoglobulin in PBS containing 0.01M sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4.
Gene Accession No. NM_153822.2; NM_002810.2
Gene Symbols PSMD4, pUB-R5, OTTHUMP00000059963, Rpn10, AF-1, AF, MCB1, S5A, ASF
Host Species Mouse
Immunogen Full length human recombinant Rpn10 protein.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.