Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-2,2,7-Trimethylguanosine Clone: K121,

Catalog No. NA02100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NA02100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NA02100UG Supplier MilliporeSigma™ Supplier No. NA02100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects 2,2,7-Trimethylguanosine Clone: K121 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation

The U small nuclear ribonucleoprotein particles (snRNPs) are present in all eukaryotes studied thus far and are involved in the splicing of RNA molecules. snRNPs are composed of several proteins and one or two RNA molecules. The RNA molecules (snRNAs) contain a unique 5' terminal cap structure containing the nucleoside 2,2,7-trimethylguanosine. The 70kDa protein component of U1 snRNP is cleaved during apoptosis, perhaps by a caspase. Monoclonal antibodies to the 5' cap structure react only to snRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 2,2,7-Trimethylguanosine
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone K121
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen 2, 7-trimethylguanosine
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the exposed 5′ cap structure (2,2,7-trimethylguanosine) of small nuclear RNAs (snRNAs).
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.