Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β2-Microglobulin Mouse anti-Human, PE, Clone: TÜ99, BD

Catalog No. BDB551337 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551337 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551337 Supplier BD Biosciences Supplier No. 551337
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

β 2-microglobulin is the 12 kDa β chain which noncovalently associates with the 44 kDa α chain to form the HLA Class I antigen complex. This antibody reacts with free or complexed β 2-microglobulin. TÜ99 does not react with β 2-microglobulin of other species. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections using TUF pretreatment.

TRUSTED_SUSTAINABILITY

Specifications

Antigen β2-Microglobulin
Applications Flow Cytometry
Classification Monoclonal
Clone TÜ99
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.