Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-20S Proteasome Core Subunits, Polyclonal

Catalog No. ST1053100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1053100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1053100UL Supplier MilliporeSigma™ Supplier No. ST1053100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects 20S Proteasome Core Subunits Clone: in Human, Mouse, Yeast samples, and it is validated for Immunohistochemistry (Paraffin), Immunoprecipitation

Proteasomes are large multi-subunit complexes, that are localized in the nucleus and cytosol, and selectively degrade intracellular proteins. They play a major role in the degradation of many proteins involved in cellular processes such as cell cycling, apoptosis, and proliferation. The human proteasome contains two copies of each of seven differentα-type andβ-type subunits assembled to form a stack of four seven-membered rings. Theα-subunits comprise the outer rings while theβ-subunits comprise the inner rings. Theβ-subunits are believed to be responsible for the proteolytic activity of the 20S proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 20S Proteasome Core Subunits
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum diluted in PBS.
Host Species Rabbit
Immunogen human erythrocyte proteasomes
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽25 to 30kDa 20S proteasome core subunits (α5,α7,β1,β5i, andβ7) in human erythrocytes, HeLa cells and 3T3 cells.
Target Species Human, Mouse, Yeast
Content And Storage −20°C
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.