Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

20S proteasome subunit α2 Mouse anti-Bovine, Human, Rabbit, Clone: MCP21, MilliporeSigma™

Catalog No. 04039MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-039-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-039-MI Supplier MilliporeSigma Supplier No. 04039
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-20S proteasome subunit α2 Antibody, clone MCP21 detects level of 20S proteasome subunit α2 and has been published and validated for use in immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 20S proteasome subunit α2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MCP21
Conjugate Unconjugated
Formulation 100μL concentration of 1mg/mL purified ascites in PBS containing 0.01M sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4.
Gene Accession No. NM_002787.3
Gene Symbols PSMA2, PSC3, MU, HC3, PSC2, PMSA2
Host Species Mouse
Immunogen Human placenta derived proteasomes
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Human, Rabbit
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.