Learn More
3-PGDH Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS571
Description
This Anti-3-PGDH Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of 3-PGDH.
Specifications
3-PGDH | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PHGDH; PGDH3 | |
KLH-conjugated linear peptide corresponding to human 3-PGDH near the N-terminus. | |
100 μg | |
NP_006614 | |
2-8°C for 12 months |